VWR® Bead Mill Homogenizer, 120 V
Supplier: VWR International
19-080
75840-022EA 8900.27 USD
75840-022
VWR® Bead Mill Homogenizer, 120 V
Homogenizers Benchtop Homogenizers
The VWR® Bead Mill is part of the newest generation of Bead Mill homogenizers. This economical system is specifically designed for grinding, lysing and homogenization of up to 16 samples. Using sample tubes pre-filled with a variety of lysing beads, the VWR® Bead Mill vigorously and uniformly shakes the tubes providing an efficient, consistent, high-yield and quality homogenization usually in less than 40 seconds.
  • 16 sample capacity
  • Run time 1 second to 10 minutes
  • 0.8 to 6 m/s
  • Up to 99 programs
  • Compatible with VWR® pre-filled bead tubes
  • TUV Certified
  • Two-year warranty
The VWR® Bead Mill simultaneously homogenizes 12×0.5, 4×1.5, 12×2, or 4×7ml samples.
The VWR® Bead Mill is an ideal solution for releasing DNA, RNA and proteins from tough samples.
It does not require a cool down between runs which allows for non-stop use and high throughput. The unit features a convenient front loading design with snap in tube carriage.
Certifications: c TUV US Certified
Ordering information: Includes the Bead Mill Drive Unit, Tube Carriage, User Manual, and a two-year warranty. Bead Mill disposable bead tubes sold seperately.