PIG® Retract-a-Clamp® Insulated Bonding/Grounding Wires, New Pig
Supplier: New Pig
DRM416
75839-192EA 336.46 USD
75839-192
PIG® Retract-a-Clamp® Insulated Bonding/Grounding Wires, New Pig
Grounders Grounding Wires
Wire creates safe grounding connection while staying coiled to avoid creating a tripping hazard.
  • Bonding and grounding assembly controls static electricity to prevent sparking while transferring or storing flammable liquids; FM approved and NFPA 77 compliant
  • Vinyl coating protects wire from moisture and corrosion for longer life and to allow outdoor use
Wire creates a solid grounding connection while staying coiled so as not to be a tripping hazard.