TSX Series High-Performance −20 °C Manual Defrost Enzyme Freezers, Thermo Scientific

Supplier: Thermo Fisher Scientific
TSX2320ED TSX2320EA TSX3020EA TSX3020ED
75839-108EA 13500 USD
75839-108 75839-116 75839-180 76320-310 76320-312 76320-314
TSX Series High-Performance −20 °C Manual Defrost Enzyme Freezers, Thermo Scientific
Freezers Laboratory / General Purpose Freezers
Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers offer a sustainable choice for enzyme storage.

  • Whisper quiet with a sound level of 52 dBA
  • V-drive technology
  • Up to 50% less energy usage than conventional refrigerant models
  • Lower HVAC costs as a result of less heat emission into the environment
  • Easy to use microprocessor control with setpoint security and alarms
  • Compatible with wireless monitoring systems and remote alarm contacts
  • Standard door locks provide comfort and assurance
  • Ergonomic door handle

Thermo Scientific™ TSX Series high-performance -20°C manual defrost enzyme freezers are designed with features that support sample protection and sustainability objectives for the storage of enzymes requiring -20°C

Units are whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction. Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection. Refrigerators offer up to 50% less energy usage than conventional refrigerant models.

Freezers feature manual defrost and microprocessor display.

Temperature range of -35° to -15°C, factory setpoint at -20°C.

Certifications: Refrigerators use natural, R290 refrigerants, making the TSX Series compliant with the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP). Manufactured in an award-winning, zero-waste to landfill facility. Refrigerators are UL, cUL certified.

Accessories information: Optional chart recorders available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR