BioClean™ Vijon™ Sterile Single Use Goggles, Ansell
Supplier: Ansell Healthcare
BVGS
75839-022CS 519.27 USD
75839-022
BioClean™ Vijon™ Sterile Single Use Goggles, Ansell
Goggles
BioClean™ Vijon™ sterile single use goggles are lightweight and non-linting.
  • Lightweight PVC frame
  • Indirect ventilation system to maintain user comfort
  • Optically correct lens
  • Toughened polycarbonate, anti-fog, and anti-scratch lens
  • Non-linting, latex-free head band offering unrivaled comfort
  • ISO Class 4 compatible
BioClean™ Vijon™ sterile single use goggles are constructed from lightweight, ultra-soft PVC. They may also be worn over eyeglasses.
Certifications: Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Conforms to Category 2 - Personal Protective Equipment PPE Directive 89/686/EEC.
Complies with requirements of EN 166:2001 standard for personal eye protection.
Tested to ANSI/ISEA Z87.1-2010, the American National Standard for Occupational and Educational Personal Eye and Face Protection Devices.
Packaging: Goggles are packaged one piece per sealed PE inner bag; 10 inner bags per sealed outer PE bag; six outer bags per lined carton (60 pieces).