SteriPlast® Spatulas for Foodstuffs, PS, Burkle
Supplier: BURKLE INC
5378-3009
75836-570PK 37.73 USD
75836-570
SteriPlast® Spatulas for Foodstuffs, PS, Burkle
Spatulas
Spatula is ideal for sampling powders, granulates and pastes.
  • Can be used as part of HACCP/IFS foreign object management
  • Individually packaged and sterilized by gamma rays
  • Especially for the food, animal feed and pharmaceutical industries
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
Certifications: Production, assembly and packaging according to cleanroom class 7. Complies with FDA regulations.