Benchtop Orbital Shaker, Eberbach
Supplier: Eberbach
E6130.00
75836-366EA 8341.5 USD
75836-366
Benchtop Orbital Shaker, Eberbach
Shakers and Mixers Orbital Shakers
This bench top variable speed orbital shaker has been designed to occupy minimal bench space and provide the versatility offered by larger units with its increased load capacity.
  • Digital tachometer and digital countown timer available
  • Motor with 35 mhp
  • Permanently lubricated bearings
  • Rated for continuous operation
The desired rate of rotation is set using the speed control knob on front panel and speed is monitored via tachometer. The digital countdown timer can be selected for timed operation of the unit, unit can also be used for constant operation.
This can be used with optional utility box, flask holders.