Headspace Crimp/Screw Top Vials, 20 mm, PerkinElmer
Supplier: PerkinElmer
N9306079 N9302134 N9306075 B0104236 N9306078 N6356478 N9303349 N9306240 N9306241 N9303348
101683-232PK 565.34 USD
101683-232 97019-614 97019-386 97019-606 97012-218 97019-612 10132-954 10136-816 10140-076 10140-078 75835-986
Headspace Crimp/Screw Top Vials, 20 mm, PerkinElmer
Vials
These headspace vials are manufactured from Type I borosilicate glass, which meets all USP, JP, and EP Pharmacopeia requirements.
  • Clear galss
  • Resistant to high temperature
  • Chemical resistant to acidic, neutral, and alkali solutions
  • Complete with pressure relief caps/septa which ensure safe analysis at any temperature