Weathered Diesel Fuel #2 Kit, Restek
Supplier: RESTEK CORP
31239
75835-626KT
155.55
USD
75835-626
Weathered Diesel Fuel #2 Kit, Restek
Standards
Petroleum Industry Standards
The kit consists of different standards.
- Contains 1 ml each of these mixtures,
- Diesel fuel #2 standard
- Diesel fuel #2 standard - 25% weathered
- Diesel fuel #2 standard - 50% weathered
- Diesel fuel #2 standard - 75% weathered
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...