CCME F1 Retention Time Marker, Restek
Supplier: RESTEK CORP
30611
75835-602EA 45.5 USD
75835-602
CCME F1 Retention Time Marker, Restek
Standards Organic Standards
The retention time marker consists of 3 components: (C6) n-hexane, (C10) n-decane, and toluene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.