Certified BTEX in Unleaded Gas Composite Standard (9 Components), Restek
Supplier: RESTEK CORP
30237
75835-548EA 41.5 USD
75835-548
Certified BTEX in Unleaded Gas Composite Standard (9 Components), Restek
Standards Organic Standards
The BTEX in unleaded gas composite standard consists of 9 components, they are benzene, ethylbenzene, isopropyl benzene, methyl tert-butyl ether (MTBE), naphthalene, toluene, m-xylene, o-xylene and p-xylene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.