Grob DA 280 Column Test Mix (11 components), Restek
Supplier: RESTEK CORP
35252
75835-376EA
53.18
USD
75835-376
Grob DA 280 Column Test Mix (11 components), Restek
Standards
Chromatography Standards
Contains: n-decane, C10, 280 µg/ml, methyl decanoate, C10:0, 420 µg/ml, n-undecane, C11, 280 µg/ml, methyl undecanoate, C11:0, 420 µg/ml, methyl dodecanoate, C12:0, 420 µg/ml, L(+)-2,3-butanediol, 530 µg/ml, 2,6-dimethylaniline, 320 µg/ml, 2,6-dimethylphenol, 320 µg/ml, 2-ethylhexanoic acid, 380 µg/ml, nonanal, 400 µg/ml, 1-octanol, 360 µg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)