NPI-1000 Column Test Mix (8 Components), Restek
Supplier: RESTEK CORP
35224
75835-360EA 49.5 USD
75835-360
NPI-1000 Column Test Mix (8 Components), Restek
Standards Chromatography Standards
The mix consists of (C9:0) methyl nonanoate, (C13) n-tridecane, (C15) n-pentadecane, acenaphthylene, 4-chlorophenol, 1,6-hexanediol, 4-propylaniline and 1-undecanol.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.