Chiral Column Test Mix (9 Components), Restek
Supplier: RESTEK CORP
35098
75835-348EA 49.5 USD
75835-348
Chiral Column Test Mix (9 Components), Restek
Standards Chromatography Standards
The mix consists of (C10:0) Methyl decanoate (210 μg/ml), (C11:0) methyl undecanoate (210 μg/ml), (C12) n-dodecane 150 μg/ml, (C12:0) methyl dodecanoate (210 μg/ml), 2,3-butanediol (+/-) (500 μg/ml), 1-heptanol (180 μg/ml), 3-methylphenol (m-cresol) (160 μg/ml), 1-phenylethanol (+/-) (500 μg/ml) and α-pinene (+/-) (500 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.