QuEChERS Internal Standard Mix for GC-MS Analysis (6 Components), Restek
Supplier: RESTEK CORP
33267
75835-214EA 43.5 USD
75835-214
QuEChERS Internal Standard Mix for GC-MS Analysis (6 Components), Restek
Standards Organic Standards
Ready to use for QuEChERS extractions—no dilutions necessary.
It consits of PCB 18 (50 μg/ml), PCB 28 (50 μg/ml), PCB 52 (50 μg/ml), triphenylmethane (10 μg/ml), triphenylphosphate (20 μg/ml), tris-(1,3-dichloroisopropyl)phosphate (50 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.