QuEChERS Internal Standard Mix for GC-NPD and LC-MS/MS Analysis (2 Components), Restek
Supplier: RESTEK CORP
33266
75835-212EA 42.5 USD
75835-212
QuEChERS Internal Standard Mix for GC-NPD and LC-MS/MS Analysis (2 Components), Restek
Standards Organic Standards
Ready to use for QuEChERS extractions—no dilutions necessary.
It consists of triphenylphosphate (20 μg/ml) and tris-(1,3-dichloroisopropyl)phosphate (50 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.