EFSA PAH4 Standard, Restek
Supplier: RESTEK CORP
32469
75835-038EA 53 USD
75835-038
EFSA PAH4 Standard, Restek
Standards Organic Standards
  • European Food Safety Authority (EFSA) PAH4 compounds prepared in toluene solvent
  • High 1,000 µg/mL concentration—dilute as needed to economically prepare custom mixes
  • Low volatility—longer shelf life and more accurate results.
  • High miscibility—compatible with QuEChERS solvents
  • Certified reference material (CRM) meets strict ISO quality requirements
  • Pair with Rxi-PAH GC column for easy separation of EFSA PAH4 compounds from common interferences in food and environmental samples: www.restek.com/rxi-pah