Chlorinated Acids by LC, Mix A (8 Components), Restek
Supplier: RESTEK CORP
32431
75834-986EA 109.52 USD
75834-986
Chlorinated Acids by LC, Mix A (8 Components), Restek
Standards Organic Standards
Consists of eight components, those are acifluorfen, bentazon, chloramben, 2,4-d, dicamba, dichlorprop, picloram and 2,4,5-tp.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.