QuEChERS Performance Standard C, Restek
Supplier: RESTEK CORP
31155
75834-630EA 116.5 USD
75834-630
QuEChERS Performance Standard C, Restek
Standards Organic Standards
  • Designed for use in all QuEChERS methods for pesticides in fruits and vegetables, including the original unbuffered method, AOAC 2007.01, and EN15662
  • Ideal for initial method evaluations and ongoing method performance validations.
  • Optimized blend of chemically compatible analytes for maximum stability and shelf life.
  • Precise formulation improves data quality and operational efficiency; spend more time running samples and less time sourcing and preparing standards
  • Quantitatively analyzed to confirm the composition and stability of each mixture
  • Produced and tested in accordance with ISO Guide 34 and 17025 accreditation.
Certifications: AOAC 2007.01 and EN15662. Produced and tested in accordance with ISO 17034 and 17025 accreditation.