QuEChERS Performance Standard A (16 Components), Restek
Supplier: RESTEK CORP
31153
75834-626EA 96 USD
75834-626
QuEChERS Performance Standard A (16 Components), Restek
Standards Organic Standards
QuEChERS performance standard A designed for use in all QuEChERS methods for pesticides in fruits and vegetables. Ideal for initial method evaluations and ongoing method performance validations.
  • Optimized blend of chemically compatible analytes for maximum stability and shelf life
  • Quantitatively analyzed to confirm the composition and stability of each mixture
Precise formulation improves data quality and operational efficiency; spend more time running samples and less time sourcing and preparing standards.
This standard contain 16 components: Acephate, Azinphos methyl, Chlorpyrifos, Coumaphos, Diazinon, Dichlofluanid, Dichlorvos, Dimethoate, Fenthion, Malathion, Methamidophos, Mevinphos, Omethoate, Phosalone, Pirimiphos methyl and Propargite.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements. It including the original unbuffered method, AOAC 2007.01, and EN15662.