508.1 Calibration Mix #3 (8 Components), Restek
Supplier: RESTEK CORP
32096
75834-226EA 58 USD
75834-226
508.1 Calibration Mix #3 (8 Components), Restek
Standards Organic Standards
Consists of eight components, those are alachlor, atrazine, chlorothalonil, cyanazine , hexachlorocyclopentadiene, metolachlor, metribuzin and simazine.
8 components
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.