HPLC Mobile Phase Storage Bottle, MicroSolv
Supplier: MicroSolv Technology Corp
58807-100-A-CS
75834-186CS 1032.52 USD
75834-186
HPLC Mobile Phase Storage Bottle, MicroSolv
Bottles HPLC Bottles
These amber borosilicate glass bottles have a 100 ml 'Total Fill Capacity' and are perfect as HPLC Mobile Phase Storage Bottles. The small openings and enamel graduations and marking patches makes them a convenient way to store concentrated Mobile Phase or its Ingredients.
Blue, solid screw caps without liners and polypropylene drip rings are included. This bottle is not 'safety coated'.