BioClean™ Clearview™ Sterile Single Use Goggles, Ansell
Supplier: Ansell Healthcare
BCGS1
75834-128CS 803.2 USD
75834-128
BioClean™ Clearview™ Sterile Single Use Goggles, Ansell
Goggles
BioClean™ Clearview™ Sterile Single Use Goggles feature an anti-fog and optically correct lens.
  • Lightweight PVC frame
  • Indirect ventilation system
  • Toughened polycarbonate, anti-scratch lens
  • Non-linting, latex-free head band
With PPE Cat 2 certification, they provide excellent product and personal protection. Goggles feature an optically correct toughened polycarbonate lens with anti-fog and anti-scratch coating, and an indirect ventilation system to maintain user comfort. They can also be worn over eyeglasses.
Certifications: Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Conforms to Category 2 – Personal Protective Equipment PPE Directive 89/686/EEC.
Complies with requirements of EN 166:2001 standard for personal eye protection.
Tested to ANSI/ISEA Z87.1-2010, the American National Standard for Occupational and Educational Personal Eye and Face Protection Devices.
Packaging: Goggles are packaged one piece per sealed Tyvek™/PE inner bag; 10 inner bags per sealed outer Tyvek™/PE bag; six outer bags per lined carton (60 pieces).