BioClean-C™ Chemotherapy Protective Sleeve Covers, Ansell
Supplier: Ansell Healthcare
S-BCSC BCSC
75834-076CS 656.65 USD
75834-076 75834-130
BioClean-C™ Chemotherapy Protective Sleeve Covers, Ansell
Disposable Garments
BioClean-C™ Chemotherapy Protective Sleeve Covers are designed for protection.
  • Tested against ASTM D6978-05 using a minimum of nine cytotoxic drugs
  • Ultrasonically bonded seams with protective tape
  • Elasticated for secure fit
  • Processed to ensure ISO Class 4 compatibility
  • 100% polyester elasticated cuffs
  • Lightweight, low-linting & hard wearing CleanTough™ material
  • ISO Class 4 compatible
Designed for protection, the BioClean-C™ Chemotherapy Protective Sleeve Covers have been specially developed for maximum protection against a range of chemotherapy drugs. They are compatible with ISO Class 4 cleanrooms. Constructed from lightweight, low-linting, and hard wearing CleanTough™ material. Sleeve covers feature elasticated arms and 100% polyester elasticated cuffs. These 'one size fits all' covers have been tailored for quick and simple donning. Sleeves are available in sterile and nonsterile formats.
Certifications: Manufactured in a facility holding ISO 9001:2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Tested against standard EN ISO 13034 protective clothing against liquid chemicals (Type PB [6]).
Tested against standard EN 14325 protective clothing against chemicals.
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Processed in an NEBB certified ISO Class 4 environment.
Packaging: Sterile sleeves are packaged one pair per sealed inner PE bag; 15 inner bags per sealed outer PE bag; six outer bags per lined carton (90 pairs). Nonsterile sleeves are packaged 15 pairs per sealed PE bag; six bags per lined carton (180 pieces).