1 kb DNA Ladder, IBI Scientific
Supplier: IBI Scientific
IB01351
75832-840EA 76.22 USD
75832-840
1 kb DNA Ladder, IBI Scientific
Electrophoresis Reagents Molecular Weight Markers
The IBI 1 kb DNA Ladder, containing 13 linear double-stranded DNA fragments is suitable for sizing double-stranded DNA from 250 to 10000 bp.
  • Ready-to-use DNA Ladder (pre-mixed with loading dye)
  • Suitable for sizing double-stranded DNA from 250 bp to 10 kb with enhanced intensities at 1 and 3 kb
  • Concentration: 2.5 µg/25 µl (Recommended loading: 0.5 µg or 5 µl)
The 1 kb DNA Ladder is a unique combination of a number of proprietary plasmids digested with appropriate restriction enzymes and PCR products to yield 13 fragments, suitable for use as molecular weight standards for electrophoresis. The enhanced intensities of the 1 and 3 kb fragments were designed for quick and easy determination of electrophoresis results.
The IBI 1 kb DNA ladder comes ready to use, no diluting necessary. Pre-mixed with 6 X loading dye made up of bromophenol blue and xylene cyanol FF.
Ships cold on blue ice and can be stored at 25 °C for 6 months, 4 °C for 12 months and −20°C for 2 years.