HardyDisks™ AST X Factor HardyDisk™, Hemin Disk, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z7031
75832-738EA 49.41 USD
75832-738
HardyDisks™ AST X Factor HardyDisk™, Hemin Disk, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
X Factor HardyDisk™, Hemin disk is designed to aid in the identification of Haemophilus spp.
Contains Hemin (X factor) for the speciation of Haemophilus. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispenser by BD BioSciences. The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP), and have achieved ISO 13485 certification as a medical device manufacturer.