HardyDisks™ AST V Factor HardyDisks™, NAD Disk, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z7041
75832-736EA 66.3 USD
75832-736
HardyDisks™ AST V Factor HardyDisks™, NAD Disk, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
V Factor HardyDisks™, NAD disk is designed to aid in the identification of Haemophilus spp.
Contains NAD (V factor) for the speciation of Haemophilus. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispenser by BD BioSciences. The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP), and have achieved ISO 13485 certification as a medical device manufacturer.