BD Biosciences Meropenem, MEM-10 Cartridge, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
231703
75832-514EA
69.01
USD
75832-514
BD Biosciences Meropenem, MEM-10 Cartridge, Hardy Diagnostics
Microbiology Test Systems
Antibiotic Tests
BBL™ Sensi-Discs™ are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The BBL™ Sensi-Disc™ cartridges are designed to be fully compatible with the BBL™ single or multiple disk dispensers.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)