VWR® Variable Speed Rocker

Supplier: VWR International
75832-308
75832-308EA 949 USD
75832-308
VWR® Variable Speed Rocker
Shakers and Mixers Rotating/Rocking Shakers
Designed to provide precise speed and tilt angles for a broad range of molecular and biological mixing applications.

  • Variable speed control up to 32 rpm
  • Easily adjustable tilt angle, from 0 to 30°
  • Large 14×12" work surface incudes non-skid rubber mat
  • Suitable for use in a cold room
  • Optional stacking tray
  • 2 year warranty

With variable speed control and adjustable tilt angles, the VWR® Rocker is ideal for many different applications including, gel staining, washing, antibody staining and hybridizations.

A large platform with a non-skid surface is included, ideal for securing all popular laboratory vessels. A powerful brushless motor drive provides smooth and consistent rocking motion and speed is adjustable to provide rocking from gentle washing to aggressive mixing.

Certifications: CE Marked.

Ordering information: Optional 14 x 12" stacking platform is available. VWR® 2D Variable Speed Rocker comes with a two-year warranty.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR