PBS Buffered Saline 1X w/o Ca & Mg, Cytiva
Supplier: Cytiva
BSS-PBS-1X6
75832-012CS 133.63 USD
75832-012
PBS Buffered Saline 1X w/o Ca & Mg, Cytiva
Buffers Electrophoresis Buffers Washing Buffers
Cytiva manufactures PBS utilizing USP raw materials formulated in USP Water for Injection quality water.
  • GMP manufactured
  • PH: 7.1 -7.3 (USP <791>)
  • Osmolality: 280 -320 mOsm/kg (USP <785>)
Phosphate-buffered saline (PBS) 1X is a physiological salt solution commonly found in many cell culture applications. PBS is isotonic and nontoxic to cells. Cytiva offers PBS that is sterile with endotoxin levels tested for each lot.