Water, Ultrapure cell & molecular grade, sterile

Supplier: Cytiva

Synonyms: Dihydrogen oxide

WPW-CMG-1X6
75832-010CS 116.7 USD
75832-010
Water, Ultrapure cell & molecular grade, sterile
Water

A universal, ultra pure water for your most sensitive research applications.


  • This product is sterile and extremely low in endotoxin
  • It is suitable for cell culture applications, media and reagent grade preparations
  • Purified utilizing a nine step process resulting in very high quality water


This water has been analyzed for the absence of nucleases and proteases for your molecular biology applications. It is suitable for research investigations of nucleic acid and protein systems.


Certifications: Manufactured under cGMP and ISO 13485 guidelines in an FDA registered facility.

Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Appearance Clear Liquid (Visual)
Sterility Sterility (USP 71)
Endotoxin 0.25 EU/mL (USP 85)
TOC (before packaging) 500 ppb (USP 643)
Conductivity 5.0 µs/cm (USP 645)
DNAse None Detected
RNAse None Detected
Protease None Detected
GMP manufactured conform

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR