Avantor® Hichrom, Guard Cartridge Holder for HPLC Columns
Supplier: Avantor
80101/N
75830-798EA 169.82 USD
75830-798
Avantor® Hichrom, Guard Cartridge Holder for HPLC Columns
Guard Column Holders
This guard cartridge holder promotes increased column lifetime.
  • Improved hardware
  • Higher pressure rating
  • Low internal swept volume
This guard cartridge holder is supplied in new hardware, leading to more robust and higher efficiency separations. Chromatographic selectivity will be unaffected by this change.