Avantor® Hichrom, Carbohydrate Cation HPLC Columns
Supplier: Avantor
70057
75830-598EA 2177.8 USD
75830-598
Avantor® Hichrom, Carbohydrate Cation HPLC Columns
Chromatography Columns
This column consists of a highly efficient sulphonated polystyrene resin supplied in the calcium form.
  • Sulphonated polystyrene resin in the calcium form
  • 100% water used as mobile phase
  • Column heating required
  • USP L19
This column provides excellent separations using only water as the mobile phase.