2-Propanol 99% antiseptic, cleanser, Hydrox Laboratories
Supplier: Healthcare Products
|
Danger
|
Synonyms:
Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol
HDX A0053
HDXD0052
75830-308CS
282.12
USD
75830-308
75830-312
2-Propanol 99% antiseptic, cleanser, Hydrox Laboratories
2-Propanol
- Topical antiseptic
- Active ingredient: isopropyl alcohol
Formula:
(CH₃)₂CHOH MW: 60.1 g/mol Melting Pt: –89 °C Flash Pt: 12 °C |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 UN: 1219 ADR: 3,II |
|
Specification Test Results
Assay | 99% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...