Tissue-Tek VIP®6 AI Vacuum Infiltration Processor, Sakura Finetek
Supplier: Sakura Finetek
6040
75818-110EA 96000.14 USD
75818-110
Tissue-Tek VIP®6 AI Vacuum Infiltration Processor, Sakura Finetek
Tissue Processors
The first and only tissue procesor offering automatic, onboard preparation of mixed reagent solutions for advanced infiltration of fatty tissues.
  • Automatic Transfer allows in-process solution rotation
  • Automatic paraffin rotation and waste container save time and prevent spills
  • 2 bulk reagent reservoirs provide fresh reagent during processing, if needed
  • Heated retort lid reduces condensation while eliminating fumes
  • Automatic bottle connection check to reduce user errors
  • User-defined cleaning cycles w/optional drying cycle to eliminate fumes
  • Illuminated reagent cabinet enables observation of reagent levels
  • Automatic paraffin degassing aspirates evaporated clearing agent from paraffin oven, keeping paraffin clean while filtering the fumes
Most reliable processing with industry leading mean time between repairs of over 52 weeks. The high-resolution LCD color touch screen and user-friendly software is capable of up to 300 cassettes per run. Options for processing include 10 user-selectable mixing modes, heat, pressure and vacuum. Tissue-Tek® iSupport™ remote system monitorying Sakura's professional technical support team ensure maximum uptime.
Certifications: UL61010-1;2nd Ed.:2004
CAN/CSA-C22.2 No.61010-1-12
IEC 61010-2-010:2003
IEC 61010-2-101:2002.
Accessories information: Accessories include two activated carbon fume filters, two 150-cassette baskets, one USB memory stick, one set of reagent bottle labels, once case of paraffin waste bags, one large paraffin scraper, one small paraffin scraper, one basket transport tray, one level sensor cleaning brush and one operating manual.