Wound Closure Tray II, Sklar
Supplier: Sklar
96-1731
75817-518CS 607.01 USD
75817-518
Wound Closure Tray II, Sklar
Trays/Dishes/Pots Instrument Trays/Dishes
Wound Closure Trays are industry standard
Each tray includes the following instruments and prep materials: Wire Formed Scissors, Plastic Dressing Forceps, Plastic Curved Hemostat, PVP Swabstick, Alcohol Swabstick, (4) Gauze Sponges (4x4", 12-ply), Absorbent Towel (17x19"), Fenestrated Drape (18x26"), Refuse Bag with Tie, and Tray.