Axygen® Agarose, Low Melting Point, Corning
Supplier: Corning
AGR-LM-100
75816-208CS 261.9 USD
75816-208 75816-210
Axygen® Agarose, Low Melting Point, Corning
Electrophoresis Gels Agarose and Acrylamide
A high quality molecular biology grade agarose suitable for analytical and preparative electrophoresis of nucleic acids.
  • Low melting temperatures and gel strength
  • Electrophoresis of DNA fragments ≥1000 bp
  • Suitable for tissue and cell culture and viral plaque assays
  • Ideal for preparative electrophoresis
A low melting agarose that allows for the recovery of undamaged nucleic acids below their denaturation temperature.
In addition, in-gel manipulations can be performed without prior extraction of the DNA from the gel slice.