Corning® Gosselin™ Petri Dish, Corning
Supplier: Corning
BP143-06
75816-186CS 172.92 USD
75816-186
Corning® Gosselin™ Petri Dish, Corning
Culture Dishes Petri Dishes
Transparent PS Petri dishes for food industry laboratories.
  • With three vents
  • Optimal flatness
  • Sterile or aseptic
  • Crystal polystyrene
These dishes are produced in a Class 100 (ISO 5) manufacturing environment, ensuring an aseptic product. Product is labelled with a batch number on each carton. Also available sterile (Sterility Assurance Level based on ISO 11137 : SAL 10⁻⁶).