TraceGOLD™ TG-35MS GC Columns
Supplier: Thermo Scientific
26094-5780
10056-550EA 641.54 USD
10056-550 10056-554 10056-556 10056-562 10056-564 10056-566 10056-580 10056-584 75814-688
TraceGOLD™ TG-35MS GC Columns
Chromatography Columns
These GC columns have a higher phenyl content for useful elution order and retention time changes.
  • Mid-polarity phase, 35% diphenyl/65% dimethyl polysiloxane
  • Equivalent to USP G42 phase
Ideal for organochlorine pesticides and herbicides, pharmaceuticals, sterols, phthalate esters etc.