Middlebrook 7H11 Selective HardyFlask™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
X28
75814-682PK 144.39 USD
75814-682
Middlebrook 7H11 Selective HardyFlask™, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
Middlebrook 7H11 Selective is used for selective isolation of Mycobacterium spp.
  • Mitchison formula
  • CAPT antibiotics
  • Selective isolation
Mitchison formula with CAPT antibiotics, for selective isolation of Mycobacterium spp.