Corn Meal Agar with Tween® 80 Deep Fill Plate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
W10
75814-670PK 72.53 USD
75814-670
Corn Meal Agar with Tween® 80 Deep Fill Plate, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Corn Meal Agar is used for the cultivation of fungi.
  • Induces chlamydospore formation by Candida spp
  • Cultivation of fungi
For the cultivation of fungi. Induces chlamydospore formation by Candida spp