Sodium chloride in saline 0,85% for dilution

Supplier: HARDY DIAGNOSTICS

Synonyms: Saline, NaCl, Sea salt, Halite

U155
75814-032PK 123.2 USD
75814-032
Sodium chloride in saline 0,85% for dilution
Sodium chloride

Saline, 0.85% is used for the preparation of isotonic suspensions and dilutions of microorganisms.


  • Filtered to minimize particulates and dead bacteria
  • Preparation of isotonic suspensions
  • Dilutions of microorganisms


For the preparation of isotonic suspensions and dilutions of microorganisms. Filtered to minimize particulates and dead bacteria.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR