GN Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
R76BX
75814-030PK 564.04 USD
75814-030
GN Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
GN Broth is used for the selective enrichment of Salmonella and Shigella spp
  • Selective enrichment of Salmonella and Shigella
For the selective enrichment of Salmonella and Shigella spp. in stool specimens.
in stool specimens.