Methylene blue (Loeffler)
Supplier: HARDY DIAGNOSTICS
Synonyms:
Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
Z88
75814-024EA
27.78
USD
75814-024
Methylene blue (Loeffler)
Methylene blue
Methylene blue, Loeffler formulation, Stain is used to examine metachromic granules in C.diphtheriae
- Bacterial and white blood cell stain
- Enumeration of fecal leukocytes
Used to examine metachromic granules in C. diphtheriae, screening bacteria in CSF, or for enumeration of fecal leukocytes.
Formula:
C₁₆H₁₈ClN₃S MW: 319.86 g/mol |
MDL Number:
MFCD00150008 CAS Number: 61-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...