HardyVal™ Tryptic Soy Broth, USP, Media-Bag™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
U100
75814-002PK
152.86
USD
75814-002
HardyVal™ Tryptic Soy Broth, USP, Media-Bag™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
Compliance made easy to verify the efficacy of aseptic technique.
- The Media-Bag™ has both a spike-port and needle-port
- Terminally autoclaved for high sterility assurance
- Hang Notch on the top of the Media Bag™ for convenience
- Packaged in double 2 ml zippered bags
- Growth promotion testing performed on each lot of media
Tryptic Soy Broth is recommended for use as a general-purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi. The U.S. Pharmacopeia National Formulary (USP) describes its use for sterility testing.
Ideal for verifying aseptic technique of pharmacists, nurses, or pharmaceutical manufacturing technicians.
Quality tested to USP 71 standards, manufactured in an ISO 13485 facility.