Vancomycin Disks, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z7501
75813-974EA 35.87 USD
75813-974
Vancomycin Disks, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
Vancomycin disks are used for the differentiation of anaerobic bacteria based on susceptibility to vancomycin.
  • Differentiation of anaerobic bacteria
  • Spring loaded for smooth and reliable dispensing
For the differentiation of anaerobic bacteria based on susceptibility to vancomycin. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispenser.