Lauryl Tryptose Broth with MUG (Durham Tube), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
K238
75813-960PK 64.04 USD
75813-960
Lauryl Tryptose Broth with MUG (Durham Tube), Hardy Diagnostics
Assays Immunological Assays
Recommended as a test of Escherichia coli and coliforms by fermentation with gas production and fluorogenic means
  • Fluorogenic test means
  • Test of Escherichia coli and coliforms
  • Gas production and MUG reaction detection
For the detection of E. coli by gas production and MUG reaction.