Zein NF

Supplier: Spectrum Chemical Mfg. Corp.

Z1131-25KGBL
75812-634EA 4538.35 USD
75812-634
Zein NF
Zein

ZEIN, NF, a class of prolamine protein found in corn, is typically manufactured as a powder from corn gluten meal and has a variety of uses as a coating for candy, nuts, fruit, pills, and other encapsulated foods and drugs. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Storage Temperature: Ambient
MDL Number: MFCD00062414
CAS Number: 9010-66-6

Order Now

Specification Test Results

Protein Content (Dried Basis) 81.9 - 100.0 %
Loss On Drying max.8.0 %
Hexane-Soluble Matter max. 12.5 %
Residue On Ignition max. 2.0 %
Heavy Metals max. 20 µg/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR