Sulfuric acid 95.0-98.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

S1736-2.5LGL-C S1736-2.5LPV-C S1736-2.5LTGL S1736-2.5LTPV S1736-200LTBL S1736-500ML-CS S1736-500MLGL
75812-378CS 1025.65 USD
75812-378 75812-380 75812-382 75812-384 75812-386 75812-388 75812-390
Sulfuric acid 95.0-98.0% FCC
Sulfuric acid

Sulfuric Acid, FCC is considered a general purpose food additive and is used to directly control pH during the processing of food by discouraging the growth of bacteria and spoilage microbes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Corrosion;Skull and crossbones
UN1830

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.840
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Assay 95.0 - 98.0 %
Arsenic (As) max. 3 mg/kg
Chloride (Cl) max. 0.005 %
Iron (Fe) max. 0.02 %
Lead (Pb) max. 5 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR