Sulfuric acid 95.0-98.0% NF
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
S1731-2.5LTP
S1731-200LTB
S1731-2.5LTG
S1731-500MLG
S1731-2.5LGL-C
S1731-2.5LPV-C
S1731-20LT
S1731-500ML-CS
700000-588EA
366.95
USD
700000-588
700000-590
700000-586
700000-592
75812-370
75812-372
75812-374
75812-376
Sulfuric acid 95.0-98.0% NF
Sulfuric acid
Sulfuric Acid, NF is used as an acidifying agent.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.840 Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 UN: 1830 ADR: 8 II |
Specification Test Results
Assay (H₂SO₄) | 95.0 - 98.0 % |
Residue On Ignition | max. 0.005 % |
Chloride (Cl) | max. 50 ppm |
Arsenic (As) | max. 1 ppm |
Heavy Metals | max. 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...