Sodium thiosulfate pentahydrate, crystals, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

S1497-12KG S1497-2.5KG S1497-500GM S1497-500GM-CS
75812-322EA 1180.35 USD
75812-322 75812-324 75812-326 75812-328
Sodium thiosulfate pentahydrate, crystals, BiotechGrade
Sodium thiosulphate pentahydrate

Sodium Thiosulfate, Pentahydrate, Crystal, BiotechGrade has been used to lessen some of the side effects of cisplatin (a cancer medicine). It is also used with another medicine in the emergency treatment of cyanide poisoning. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.


Spectrum offers highly pure reagents suitable for biochemical research and analysis.

Formula: Na₂S₂O₃.5H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7

Order Now

Specification Test Results

Appearance Colorless Crysta
Odor Odorless
Assay (Na₂S₂O₃) 99.0-100.5 %
Water 32.0- 37.0 %
Appearance of a 10% Solution Clear & Colorle


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR